ENA332672871 Enamine

5-bromo-3,3-diethyl-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD13192264
  • Cas Number : 304876-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79337

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79337
Availability details for
Quantity
  • Availability
  • Add to Cart