ENA332672877 Enamine

N1-ethylbenzene-1,4-diamine

  • MDL number MFCD11186960

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80358
Availability details for
Quantity
  • Availability
  • Add to Cart