ENA332672885 Enamine

3-(quinolin-4-yl)prop-2-enoic acid

  • MDL number MFCD00137153
  • Cas Number : 13026-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256...258
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80379

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80379
Availability details for
Quantity
  • Availability
  • Add to Cart