ENA332672890 Enamine

[4-(phenylamino)piperidin-4-yl]methanol

  • MDL number MFCD09955354
  • Cas Number : 113231-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80386
Availability details for
Quantity
  • Availability
  • Add to Cart