ENA332672895 Enamine

3-tert-butyl-6-hydrazinylpyridazine

  • MDL number MFCD16169000
  • Cas Number : 99565-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80392
Availability details for
Quantity
  • Availability
  • Add to Cart