ENA332673027 Enamine

4-carbamoylfuran-2-carboxylic acid

  • MDL number MFCD19382228
  • Cas Number : 1315367-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80431
Availability details for
Quantity
  • Availability
  • Add to Cart