ENA332673044 Enamine

bis(N''-(2-hydroxyethyl)guanidine); sulfuric acid

  • MDL number MFCD19382245
  • Cas Number : 1876-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2H2SO4
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80469
Availability details for
Quantity
  • Availability
  • Add to Cart