ENA332673051 Enamine

2-(thiophen-2-ylmethyl)azepane hydrochloride

  • MDL number MFCD19382252
  • Cas Number : 1315367-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80529

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80529
Availability details for
Quantity
  • Availability
  • Add to Cart