ENA332673060 Enamine

(2-tert-butyloxan-3-yl)methanamine

  • MDL number MFCD19382261
  • Cas Number : 1315365-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80572
Availability details for
Quantity
  • Availability
  • Add to Cart