ENA332673062 Enamine

N-(1,1-dioxo-1lambda6-thiolan-3-yl)-2-(piperazin-1-yl)acetamide dihydrochloride

  • MDL number MFCD19382263
  • Cas Number : 1315369-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80580
Availability details for
Quantity
  • Availability
  • Add to Cart