ENA332673074 Enamine

2-phenylcyclobutan-1-ol

  • MDL number MFCD19382278
  • Cas Number : 92243-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80622
Availability details for
Quantity
  • Availability
  • Add to Cart