ENA332673089 Enamine

2-amino-2-(2-fluorophenyl)acetamide hydrochloride

  • MDL number MFCD19382294
  • Cas Number : 1315365-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80738

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80738
Availability details for
Quantity
  • Availability
  • Add to Cart