ENA332673108 Enamine

3-bromoquinoline-8-carboxylic acid

  • MDL number MFCD19382315
  • Cas Number : 1315366-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80890

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80890
Availability details for
Quantity
  • Availability
  • Add to Cart