ENA332673118 Enamine

3-(5-methylfuran-2-yl)-1lambda6-thiomorpholine-1,1-dione

  • MDL number MFCD19382325
  • Cas Number : 1315367-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80917
Availability details for
Quantity
  • Availability
  • Add to Cart