ENA332673149 Enamine

N-(2-hydroxyethyl)pyridine-3-carboxamide

  • MDL number MFCD00547113
  • Cas Number : 6265-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80598
Availability details for
Quantity
  • Availability
  • Add to Cart