ENA332673156 Enamine

2-(3-bromophenoxy)ethan-1-ol

  • MDL number MFCD11147862
  • Cas Number : 79906-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80609
Availability details for
Quantity
  • Availability
  • Add to Cart