ENA332673157 Enamine

methyl 3-[(6-methylpyridin-2-yl)amino]propanoate

  • MDL number MFCD12806956
  • Cas Number : 103392-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80610
Availability details for
Quantity
  • Availability
  • Add to Cart