ENA332673168 Enamine

5-(cyclopropylmethyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD16665522
  • Cas Number : 302842-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80479

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80479
Availability details for
Quantity
  • Availability
  • Add to Cart