ENA332673170 Enamine

5,6-dimethoxy-1H-indole-2-carboxylic acid

  • MDL number MFCD00015459
  • Cas Number : 88210-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80483

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-80483
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart