ENA332673172 Enamine

4-[2-(trifluoromethyl)phenyl]-1,3-thiazol-2-amine

  • MDL number MFCD02664027
  • Cas Number : 383129-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80432

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80432
Availability details for
Quantity
  • Availability
  • Add to Cart