ENA332673174 Enamine

1-(bromomethyl)-2-(difluoromethoxy)benzene

  • MDL number MFCD00190103
  • Cas Number : 85684-64-6
  • Molecular Weight : 237.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 108
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-80433

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-80433
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart