ENA332673191 Enamine

2-(4-methoxyphenyl)-5-methyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD07379215
  • Cas Number : 2940-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80658
Availability details for
Quantity
  • Availability
  • Add to Cart