ENA332673192 Enamine

2-(2-chloro-4-nitrophenyl)acetic acid

  • MDL number MFCD11036396
  • Cas Number : 73088-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80502
Availability details for
Quantity
  • Availability
  • Add to Cart