ENA332673195 Enamine

ethyl 4-amino-1H-imidazole-5-carboxylate

  • MDL number MFCD00823435
  • Cas Number : 21190-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80662

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80662
Availability details for
Quantity
  • Availability
  • Add to Cart