ENA332673197 Enamine

5-methoxy-3-methyl-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD01859676
  • Cas Number : 21243-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231-234
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80503
Availability details for
Quantity
  • Availability
  • Add to Cart