ENA332673213 Enamine

6-tert-butylpyridazin-3-amine

  • MDL number MFCD17216104
  • Cas Number : 82560-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-139
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80691

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P305+P351+P338,P330,P337+P313,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-80691
pictograms GHS07
hstatements H302,H319
Availability details for
Quantity
  • Availability
  • Add to Cart