ENA332673228 Enamine

3-(ethanesulfonyl)aniline

  • MDL number MFCD11189664
  • Cas Number : 83164-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80734
Availability details for
Quantity
  • Availability
  • Add to Cart