ENA332673229 Enamine

methyl 2-(2-phenylacetamido)benzoate

  • MDL number MFCD00032216
  • Cas Number : 75541-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80737
Availability details for
Quantity
  • Availability
  • Add to Cart