ENA332673231 Enamine

4-[(methylthio)methyl]pyridin-2-amine

  • MDL number MFCD16765791
  • Cas Number : 179554-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80740

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80740
Availability details for
Quantity
  • Availability
  • Add to Cart