ENA332673236 Enamine

{3-[2-(piperidin-1-yl)ethoxy]phenyl}boronic acid

  • MDL number MFCD11540656
  • Cas Number : 1311166-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80747
Availability details for
Quantity
  • Availability
  • Add to Cart