ENA332673265 Enamine

1-(2-bromophenyl)-2-(2-fluorophenyl)ethan-1-one

  • MDL number MFCD02260756
  • Cas Number : 898784-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80798
Availability details for
Quantity
  • Availability
  • Add to Cart