ENA332673267 Enamine

1-chloro-2-(2-isocyanatoethyl)benzene

  • MDL number MFCD16302192
  • Cas Number : 93489-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80801
Availability details for
Quantity
  • Availability
  • Add to Cart