ENA332673268 Enamine

2-cyclobutoxybenzoic acid

  • MDL number MFCD16692375
  • Cas Number : 1275723-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80803
Availability details for
Quantity
  • Availability
  • Add to Cart