ENA332673277 Enamine

(2,6-difluorophenyl)hydrazine hydrochloride

  • MDL number MFCD03094169
  • Cas Number : 502496-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177...179
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80819

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80819
Availability details for
Quantity
  • Availability
  • Add to Cart