ENA332673278 Enamine

3-(trifluoromethyl)piperidin-3-ol

  • MDL number MFCD16990769
  • Cas Number : 1052713-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80830

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80830
Availability details for
Quantity
  • Availability
  • Add to Cart