ENA332673280 Enamine

thieno[3,2-b]pyridine-5-carboxylic acid

  • MDL number MFCD01927157
  • Cas Number : 56473-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80868
Availability details for
Quantity
  • Availability
  • Add to Cart