ENA332673287 Enamine

2-amino-1,3-thiazole-4-carboxamide

  • MDL number MFCD08689593
  • Cas Number : 118452-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-211
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80524
Availability details for
Quantity
  • Availability
  • Add to Cart