ENA332673302 Enamine

4-(3-aminopropyl)phenol hydrobromide

  • MDL number MFCD19380075
  • Cas Number : 583825-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80548
Availability details for
Quantity
  • Availability
  • Add to Cart