ENA332673337 Enamine

1-[1-(propan-2-yl)-1H-1,3-benzodiazol-2-yl]ethan-1-amine dihydrochloride

  • MDL number MFCD11506441
  • Cas Number : 1185299-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80587
Availability details for
Quantity
  • Availability
  • Add to Cart