ENA333557015 Enamine

2-chloro-N,N-dimethyl-6-(trifluoromethyl)pyrimidin-4-amine

  • MDL number MFCD19686205
  • Cas Number : 1334148-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27755
Availability details for
Quantity
  • Availability
  • Add to Cart