ENA333557017 Enamine

2-chloro-N-[(4-fluorophenyl)(phenyl)methyl]-N-methylacetamide

  • MDL number MFCD19686206
  • Cas Number : 1334148-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28469
Availability details for
Quantity
  • Availability
  • Add to Cart