ENA333557030 Enamine

1-{[1-(propan-2-yl)-1H-imidazol-2-yl]methyl}-1,4-diazepane

  • MDL number MFCD19686212
  • Cas Number : 1334147-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 91
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71060
Availability details for
Quantity
  • Availability
  • Add to Cart