ENA333557032 Enamine

3-ethyl-1-[2-(1H-pyrazol-1-yl)ethyl]piperazine

  • MDL number MFCD19686213
  • Cas Number : 1334146-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71130

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-71130
pictograms GHS05,GHS07
hstatements H302,H312,H318,H332
Availability details for
Quantity
  • Availability
  • Add to Cart