ENA333557033 Enamine

1-ethoxy-3-(piperazin-1-yl)propan-2-ol

  • MDL number MFCD19597916
  • Cas Number : 54469-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71175
Availability details for
Quantity
  • Availability
  • Add to Cart