ENA333557040 Enamine

octahydro-2H-1lambda6,4-benzothiazine-1,1-dione

  • MDL number MFCD19686218
  • Cas Number : 1334147-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75219
Availability details for
Quantity
  • Availability
  • Add to Cart