ENA333557041 Enamine

1-methyl-4-{5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazin-3-yl}-1H-pyrazole

  • MDL number MFCD19982341
  • Cas Number : 1334148-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75539

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75539
Availability details for
Quantity
  • Availability
  • Add to Cart