ENA333557044 Enamine

2-[(2,2,2-trifluoro-1-phenylethyl)amino]acetic acid

  • MDL number MFCD19686222
  • Cas Number : 1334146-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75921
Availability details for
Quantity
  • Availability
  • Add to Cart