ENA333557045 Enamine

[2-(furan-2-yl)-1,3-oxazol-5-yl]methanol

  • MDL number MFCD19686223
  • Cas Number : 1334148-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76252

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76252
Availability details for
Quantity
  • Availability
  • Add to Cart