ENA333557048 Enamine

5-(1H-pyrazol-1-yl)pyrimidine

  • MDL number MFCD19686224
  • Cas Number : 934838-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77009
Availability details for
Quantity
  • Availability
  • Add to Cart