ENA333557052 Enamine

4-methylpiperazine-2-carbonitrile

  • MDL number MFCD19982344
  • Cas Number : 1334146-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77749
Availability details for
Quantity
  • Availability
  • Add to Cart